missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRBOX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109893
This item is not returnable.
View return policy
Description
Immunogen sequence: MAMSQESLTF KDVFVDFTLE EWQQLDSAQK NLYRDVMLEN YSHLVSVGYL VAKPDVIFRL GPGEESWMAD GGTPVRTCAG EDRPDVSIFA SCILKCCY.
This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011].
Specifications
| KRBOX4 | |
| Polyclonal | |
| Unconjugated | |
| KRBOX4 | |
| KRAB box domain containing 4; KRAB box domain-containing protein 4; KRAB domain-containing protein 4; KRBOX4; putative zinc finger transcription factor; zinc finger family member 673; zinc finger protein 673; ZNF673 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55634 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JUW0 | |
| KRBOX4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human KRBOX4 (NP_001123371.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction