missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KRBOX4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16391635
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16391635 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16391635 Supplier Invitrogen™ Supplier No. PA584688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VDEQIDHYKE SQDKLPWQAA FIGKETLKDE SGQ Highest antigen sequence indentity to the following orthologs: Mouse - 36%, Rat - 36%.

This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRBOX4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRBOX4
Gene Accession No. Q5JUW0
Gene Alias KRAB box domain containing 4; KRAB box domain-containing protein 4; KRAB domain-containing protein 4; KRBOX4; putative zinc finger transcription factor; zinc finger family member 673; zinc finger protein 673; ZNF673
Gene Symbols KRBOX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRBOX4. Recombinant protein control fragment (Product #RP-105351).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.