missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kresoxim-methyl Monoclonal Antibody (2G4)
GREENER_CHOICE

Product Code. 17919002
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919002 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919002 Supplier Invitrogen™ Supplier No. MA549682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Kresoxim-methyl is a broad-spectrum fungicide that belongs to the group of strobilurins. It is used to control various fungal diseases in crops such as fruits, vegetables, cereals, and ornamental plants. Kresoxim-methyl works by inhibiting the respiration process of the fungi, preventing their growth and reproduction. It is highly effective against powdery mildew, rust, and leaf spot diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kresoxim-methyl
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2G4
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of KSM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.