missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRIT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19307
This item is not returnable.
View return policy
Description
KRIT1 Polyclonal specifically detects KRIT1 in Human samples. It is validated for Western Blot, Electron Microscopy.
Specifications
| KRIT1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing | |
| Rabbit | |
| 84 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (94%), Chicken (93%), Rat (95%), Bovine (98%), Xenopus tropicalis (88%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunoassay | |
| 0.74mg/mL | |
| Western Blot 1:500-1:3000, Electron Microscopy 1:10 - 1:500 | |
| O00522 | |
| KRIT1 | |
| Recombinant protein encompassing a sequence within the center region of human KRIT1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 889 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction