Learn More
KRN5, MedChemExpress
Product Code. 16487287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487287 5 mg 5mg
16473136 25 mg 25mg
16455426 100 mg 100mg
16440877 50 mg 50mg
16424578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16487287 Supplier Medchem Express Supplier No. HY112126
MedChemExpress KRN5, a derivative of KRN2, is an oral active Nuclear factor of activated T cells 5 (NFAT5) suppressor, with an IC50 of 750 nM. KRN5 has potential to treat NFAT5-mediated Chronic Arthritis.
KRN5, a derivative of KRN2, is an oral active Nuclear factor of activated T cells 5 (NFAT5) suppressor, with an IC50 of 750 nM. KRN5 has potential to treat NFAT5-mediated Chronic Arthritis.
Specifications
Chemical Name or Material KRN5
CAS 1800465-47-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H22FNO5
Quantity 5 mg
Solubility Information DMSO : 5 mg/mL (10.88 mM; ultrasonic and warming and heat to 80°C)
SMILES COC1=C(OC)C2=C(C=C1)C(CC3=CC=CC=C3F)=C4N(C2=O)CCC(C4=C5)=CC6=C5OCO6
Molecular Weight (g/mol) 459.47
Formula Weight 459.47
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only