missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22252
This item is not returnable.
View return policy
Description
KRR1 Polyclonal specifically detects KRR1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| KRR1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| HIV-1 Rev binding protein 2, HIV-1 Rev-binding protein 2, HRB2, KRR1 small subunit processome component homolog, KRR1, small subunit (SSU) processome component, homolog (yeast), Rev interacting protein, Rev-interacting protein 1, Rip-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11103 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_008974.5 | |
| KRR1 | |
| The immunogen this antibody was made to, maps to a region between residue 300 and 350 of human Small Subunit (SSU) Processome Component using the numbering given in entry NP_008974.5 (GeneID 11103). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction