missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT1, Mouse anti-Human, Clone: SPM115 + SPM116, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15226.100ug
This item is not returnable.
View return policy
Specifications
| KRT1 | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| CK1/EHK1/K1/KRT1A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| SPM115 + SPM116 | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| keratin 1 | |
| KRT1 | |
| Human epidermal keratin. | |
| RUO | |
| 3848/3850/3851/3852/3853/3854/286887/3856/3858/386 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction