missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT10/KRT13 Mouse anti-Human, Clone: DE-K13, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14967.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human KRT10/KRT13.
- High specificity
- High reactivity
Specifications
| KRT10/KRT13 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.25-0.5 ug/mL) The optimal working dilution should | |
| P13645;P13646 | |
| KRT10 | |
| Cytoskeletal preparation extracted from human ectocervical epithelium. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| DE-K13 | |
| Mouse monoclonal antibody raised against native human KRT10/KRT13. | |
| In 10 mM PBS. | |
| CK10/K10/KPP | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3858/3860 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction