missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT16 Monoclonal Antibody (OTI1G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808398S
This item is not returnable.
View return policy
Description
KRT16 Monoclonal antibody specifically detects KRT16 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| KRT16 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15211 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5863 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G5 | |
| Unconjugated | |
| RGL2 | |
| AI324768; CK16; CK-16; cytokeratin 16; cytokeratin-16; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; K16; K1CP; Ka16; keratin 16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 16; Keratin-16; Krt1-16; Krt16; KRT16A; NEPPK; PC1; type I keratin KA16 | |
| Human recombinant protein fragment corresponding to amino acids 547-777 of human RGL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction