missing translation for 'onlineSavingsMsg'
Learn More

KRT16 Monoclonal Antibody (OTI2H8), TrueMAB™, OriGene

Product Code. 17529483
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17529483 30 μL 30µL
17509483 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17529483 Supplier Origene Technologies Supplier No. TA808337S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KRT16 Monoclonal antibody specifically detects KRT16 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen KRT16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KLK3
Gene Accession No. P07288
Gene Alias AI324768; CK16; CK-16; cytokeratin 16; cytokeratin-16; FNEPPK; focal non-epidermolytic palmoplantar keratoderma; K16; K1CP; Ka16; keratin 16; keratin 16, type I; keratin complex 1, acidic, gene 16; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 16; Keratin-16; Krt1-16; Krt16; KRT16A; NEPPK; PC1; type I keratin KA16
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 25-261 of human KLK3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 354
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.