Learn More
KRT16 Monoclonal Antibody (OTI1G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808398
Encompass_Preferred
 View more versions of this product
Product Code. 17569603
  • € 540.87 / 100µL
Description
KRT16 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region of chromosome 17q12-q21. This keratin has been coexpressed with keratin 14 in a number of epithelial tissues, including esophagus, tongue, and hair follicles. Mutations in this gene are associated with type 1 pachyonychia congenita, non-epidermolytic palmoplantar keratoderma and unilateral palmoplantar verrucous nevus.
Specifications
Specifications
KRT16
Monoclonal
Unconjugated
KRT16
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
Mouse
Affinity Chromatography
RUO
3868
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
OTI1G5
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P08779
KRT16
Full length human recombit protein of human KRT16 produced in E.coli.