missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT19 Monoclonal Antibody (OTI6A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803863S
This item is not returnable.
View return policy
Description
KRT19 Monoclonal antibody specifically detects KRT19 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| KRT19 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08727 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3880 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6A8 | |
| Unconjugated | |
| KRT19 | |
| 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 | |
| Human recombinant protein fragment corresponding to amino acids 240-390 of human KRT19 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction