missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT19 Mouse anti-Canine, Human, Rat, Clone: OTI3F8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500212
This item is not returnable.
View return policy
Description
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.Specifications
| KRT19 | |
| Monoclonal | |
| 0.15 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08727, Q63279 | |
| KRT19 | |
| Recombit fragment expressed in E.coli corresponding to amino acids 240-390 of human CK19 | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F8 | |
| Unconjugated | |
| KRT19 | |
| CK19, K19, K1CS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100685481, 360626, 3880 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion