Learn More
KRT20 Monoclonal Antibody (OTI5F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802664S
This item is not returnable.
View return policy
Description
KRT20 Monoclonal antibody specifically detects KRT20 in Human samples. It is validated for Western BlotSpecifications
| KRT20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35900 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI5F1 | |
| Unconjugated | |
| KRT20 | |
| 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
| Full length human recombinant protein of human KRT20 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |