missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCRLB Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809247S
This item is not returnable.
View return policy
Description
FCRLB Monoclonal antibody specifically detects FCRLB in Human samples. It is validated for Western BlotSpecifications
| FCRLB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6BAA4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 127943 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| FCRLB | |
| fc receptor homolog expressed in B cells protein 2; Fc receptor homolog expressed in B-cells protein 2; Fc receptor like B; Fc receptor related Y; fc receptor-like and mucin-like protein 2; Fc receptor-like B; Fc receptor-like mucin-like 2; fc receptor-like protein 2; Fc receptor-related protein Y; FCRL2; FCRL2FCRLM2; FCRLB; FCRLM2; FCRLY; FcRY; FLJ31052; FREB2; FREB-2; FREB-2FcRY; mFCRL2; RGD1563018; RP11-474I16.6 | |
| Human recombinant protein fragment corresponding to amino acids 202-426 of human FCRLB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction