missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT20 Monoclonal Antibody (OTI6G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802575S
This item is not returnable.
View return policy
Description
KRT20 Monoclonal antibody specifically detects KRT20 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| KRT20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11388 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7153 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G4 | |
| Unconjugated | |
| TOP2A | |
| 9030623C06Rik; CD20; CK20; CK-20; CK-21; cytokeratin 20; cytokeratin 21; cytokeratin-20; Cytokeratin-21; K20; keratin 20; keratin 20, type I; keratin, type I cytoskeletal 20; Keratin-20; krt20; Krt21; MGC35423; Protein IT | |
| Human recombinant protein fragment corresponding to amino acids 1100-1531 of human TOP2A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction