missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT20 Monoclonal Antibody (OTI5F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802664
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.Specifications
| KRT20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35900 | |
| KRT20 | |
| Full length human recombit protein of human KRT20 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI5F1 | |
| Unconjugated | |
| KRT20 | |
| CD20, CK-20, CK20, K20, KRT21 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54474 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction