missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Krt222 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551187
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total KRT222 protein.
KRT222, also known as KRT222P. KRT222 is a 295 amino acid protein belonging to the intermediate filament family. The gene encoding KRT222P has been listed as a pseudogene, however it has not been established that the protein is not translated, and is therefore treated as a protein coding gene. Existing as two alternatively spliced isoforms, the gene encoding KRT222P maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1. Tumor suppressor p53 is necessary for maintenance of cellular genetic integrity by moderating cell fate through DNA repair versus cell death. Malfunction or loss of p53 expression is associated with malignant cell growth and Li-Fraumeni syndrome. Like p53, BRCA1 is directly involved in DNA repair, though specifically it is recognized as a genetic determinant of early onset breast cancer and predisposition to cancers of the ovary, colon, prostate gland and fallopian tubes.
Specifications
| Krt222 | |
| Polyclonal | |
| Unconjugated | |
| Krt222 | |
| 6330509G02Rik; KA21; keratin 222; keratin 222 pseudogene; keratin 222, type II; keratin-222; Keratin-222 pseudogene; keratin-like protein KRT222; Krt222; KRT222P | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 125113 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.3 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8N1A0 | |
| Krt222 | |
| Full length fusion protein of human KRT222. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction