missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT23 Antibody (OTI3F2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71780AF488
This item is not returnable.
View return policy
Description
KRT23 Monoclonal specifically detects KRT23 in Human samples. It is validated for Western Blot.
Specifications
| KRT23 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KRT23 | |
| Recombinant protein fragment corresponding to amino acids 1-270 of human KRT23 (NP_056330) produced in E.coli. | |
| 0.1 mL | |
| 25984.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI3F2 | |
| Western Blot | |
| CK23, CK-23, cytokeratin 23, Cytokeratin-23, DKFZp434G032, HAIK1, hyperacetylation-inducible type I keratin, K23histone deacetylase inducible keratin 23, keratin 23 (histone deacetylase inducible), keratin, type I cytoskeletal 23, keratin-23, MGC26158, type I intermediate filament cytokeratin | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction