missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT24 Monoclonal Antibody (OTI4E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803131
This item is not returnable.
View return policy
Description
KRT24 encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21.Specifications
| KRT24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q2M2I5 | |
| KRT24 | |
| Human recombit protein fragment corresponding to amino acids 139-397 of human KRT24 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E8 | |
| Unconjugated | |
| KRT24 | |
| K24, KA24 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 192666 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction