missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT24 Monoclonal Antibody (OTI4E8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803131S
This item is not returnable.
View return policy
Description
KRT24 Monoclonal antibody specifically detects KRT24 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| KRT24 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42771 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1029 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E8 | |
| Unconjugated | |
| CDKN2A | |
| CK-24; Cytokeratin-24; K24; KA24; keratin 24; keratin 24, type I; keratin, type I cytoskeletal 24; Keratin-24; KRT24; Type I keratin-24 | |
| Full length human recombinant protein of human CDKN2A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction