missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT35 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811831S
This item is not returnable.
View return policy
Description
KRT35 Monoclonal antibody specifically detects KRT35 in Human samples. It is validated for Western BlotSpecifications
| KRT35 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HBG7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1A4 | |
| Unconjugated | |
| Ly9 | |
| AI662400; Ha5; Ha-5; hair keratin; hair keratin acidic 5; hair keratin, type I Ha5; hard keratin, type I, 5; HHA5; HHa5 hair keratin type I intermediate filament; HKA5; K35; Ka30; keratin 35; keratin 35, type I; keratin complex 1, acidic, gene 2; keratin complex 1, acidic, gene 24; keratin complex 1, gene 24; keratin, hair, acidic, 5; keratin, type I cuticular Ha5; Keratin-35; KRT hHa5; Krt1-2; Krt1-24; KRT35; KRTHA5; type I hair keratin KA30; Unknown (protein for MGC:139866); wool (hair) keratin intermediate filament protein | |
| Human recombinant protein fragment corresponding to amino acids 48-454 of human LY9 produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction