missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT35 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811831
This item is not returnable.
View return policy
Description
KRT35 is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.Specifications
| KRT35 | |
| Monoclonal | |
| Unconjugated | |
| KRT35 | |
| HA5, Ha-5, KRTHA5, hHa5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3886 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1A4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92764 | |
| KRT35 | |
| Full length human recombit protein of human KRT35 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction