missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KRT38 Polyclonal Antibody

Product Code. 16590732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16590732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16590732 Supplier Invitrogen™ Supplier No. PA597909

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRT38 Polyclonal Antibody for Western Blot, ELISA

There are two types of hair/microfibrillar keratin, I (acidic) and II (neutral to basic).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT38
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRT38
Gene Accession No. O76015
Gene Alias HA8; hair keratin; Hair keratin, type I Ha8; HHA8; HKA8; K38; keratin 38; keratin 38, type I; keratin, hair, acidic, 8; keratin, type I cuticular Ha8; Keratin-38; KRT38; KRTHA8
Gene Symbols KRT38
Host Species Rabbit
Immunogen Recombinant Human Keratin, type I cuticular Ha8 protein (2-456AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8687
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.