missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT5 Monoclonal Antibody (OTI4A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801092
This item is not returnable.
View return policy
Description
The protein encoded by the KRT5 gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
| KRT5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P13647 | |
| KRT5 | |
| Human recombit protein fragment corresponding to amino acids 1-273 of human KRT5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4A7 | |
| Unconjugated | |
| KRT5 | |
| CK5, DDD, DDD1, EBS2, K5, KRT5A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3852 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction