missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT7 Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803076S
This item is not returnable.
View return policy
Description
KRT7 Monoclonal antibody specifically detects KRT7 in Human samples. It is validated for Western BlotSpecifications
| KRT7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P42771 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1029 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| CDKN2A | |
| CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729} | |
| Full length human recombinant protein of human CDKN2A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction