missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT7 Rabbit anti-Human, Clone: RM284, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14931.100uL
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against synthetic peptide of human KRT7.
- High specificity
- High reactivity
Specifications
| KRT7 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:1000) Western Blotting (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| P08729 | |
| KRT7 | |
| A synthetic peptide corresponding to C-terminus of human KRT7. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| RM284 | |
| Rabbit monoclonal antibody raised against synthetic peptide of human KRT7. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| CK7/K2C7/K7/MGC129731/MGC3625/SCL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3855 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction