missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KRT72 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17190774 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17190774 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190774 Supplier Invitrogen™ Supplier No. PA5112918

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRT72 Polyclonal Antibody for Western Blot, ELISA

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells. The type II keratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This gene encodes a type II keratin that is specifically expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q12-q13. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT72
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRT72
Gene Accession No. Q14CN4
Gene Alias CK-72; cytokeratin-72; K6irs; K6IRS2; K72; KB35; keratin 6, inner root sheath, 2; keratin 72; keratin 72, type II; keratin protein K6irs; keratin, type II cytoskeletal 72; Keratin-72; KRT6; KRT6IRS2; KRT72; Type II inner root sheath-specific keratin-K6irs2; type-II keratin Kb35
Gene Symbols KRT72
Host Species Rabbit
Immunogen Recombinant Human Keratin, type II cytoskeletal 72 protein (301-511AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140807
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.