Learn More
Invitrogen™ KRT76 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA546516
Product Code. 15557192
  • € 520.00 / 100µL
Description
Peptide sequence: GSISVSHSGM GSSSGSIQTS GGSGYKSGGG GSTSIRFSQT TSSSQHSSTK Sequence homology: Dog: 92%; Horse: 100%; Human: 100%; Mouse: 83%; Rat: 83%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KRT76
Polyclonal
Unconjugated
KRT76
2310001L23Rik; CK-2P; cytokeratin 2; cytokeratin-2P; HUMCYT2A; K2P; K76; Kb9; keratin 2p; keratin 76; keratin 76, type II; keratin, type II cytoskeletal 2 oral; Keratin-76; KRT2B; KRT2P; KRT76; masticatory epithelia keratin 2p; type II keratin Kb9; type-II keratin Kb9
Rabbit
Affinity chromatography
RUO
51350
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q01546
KRT76
Synthetic peptide directed towards the C-terminal of human KRT76 (aa 589-638).