missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT80 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567483
This item is not returnable.
View return policy
Description
Immunogen sequence: GEEGRMDSPSA TVVSAVQSRC KTAASRSGLS KAPSRKKKGS KGPVIKITEM SEKYFSQESE VSE Highest antigen sequence identity to the following orthologs - mouse 75%, rat 81%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene's expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.
Specifications
| KRT80 | |
| Polyclonal | |
| Unconjugated | |
| KRT80 | |
| 1200016G03Rik; 2310041I20Rik; CK-80; cytokeratin-80; K80; Kb2; KB20; keratin 80; keratin 80, type II; keratin b20; keratin, type II cytoskeletal 80; Keratin-80; KRT80; type II keratin; type II keratin Kb20; type II keratin-20; type-II keratin Kb20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 144501 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6KB66 | |
| KRT80 | |
| Recombinant Human KRT80. Recombinant protein control fragment (Product #RP-108136). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction