Learn More
KRT84 Mouse anti-Human, Clone: OTI2D1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811189
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Keratin multigene family is made of "soft" epithelial cytokeratins and "hard" hair Keratins. While the epithelial cytokeratins are involved in the layering and formation of epithelia, the hair Keratins are responsible for creating nails and hair. There are two types of Keratins: the acidic class I Keratin proteins and the basic/neutral class II Keratin proteins. Keratin 84, also known as KRT84, HB4 or KRTHB4, is a 600 amino acid protein that is expressed in hair follicles. Keratin 84 is a basic/neutral type II Keratin protein which heterodimerizes with type I Keratins to create hair and nails. The gene encoding Keratin 84 maps to human chromosome 12 which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis and Kniest dysplasia.Specifications
| KRT84 | |
| Monoclonal | |
| Unconjugated | |
| KRT84 | |
| HB4, KRTHB4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3890 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| OTI2D1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NSB2 | |
| KRT84 | |
| Full length human recombit protein of human KRT84 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.