missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT87P Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572087
This item is not returnable.
View return policy
Description
KRT87P Polyclonal Antibody for Western Blot
KRT87P (Keratin 87 Pseudogene) is a Pseudogene. Gene Ontology (GO) annotations related to this gene includestructural molecule activity.
Specifications
| KRT87P | |
| Polyclonal | |
| Unconjugated | |
| KRT87P | |
| HBD; keratin 87 pseudogene; KRT121P; KRT87P; KRTHBP4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 85349 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.89 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NCN2 | |
| KRT87P | |
| KLH conjugated synthetic peptide between 96-122 amino acids from the N-terminal region of human KT81L. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction