missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT9 Monoclonal Antibody (8C2A6)

KRT9 Monoclonal Antibody (8C2A6)
Brand: Invitrogen MA548532
This item is not returnable.
View return policy
Description
KRT9 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA
Specifications
| KRT9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P35527, Q6RHW0, Q8CIS9 | |
| KRT9 | |
| Purified recombinant fragment of human KRT9 (AA: 153-460) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 8C2A6 | |
| Unconjugated | |
| KRT9 | |
| CK-9; cytokeratin 9; cytokeratin-9; EPPK; K9; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; keratin, type I cytoskeletal 9; Keratin-9; Krt1-9; Krt9; spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9 | |
| Mouse | |
| Protein G | |
| RUO | |
| 107656, 266717, 3857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction