missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTAP2 Mouse anti-Human, Clone: OTI3B4, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503766S
This item is not returnable.
View return policy
Description
KRTAP2 Monoclonal antibody specifically detects KRTAP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| KRTAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BYR9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 85294 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3B4 | |
| Unconjugated | |
| KRTAP2-4 | |
| high sulfur keratin-associated protein 2.4; KAP2.1B; KAP2.3; KAP2.4; keratin associated protein 2.1B; keratin associated protein 2-4; Keratin-associated protein 2.3; Keratin-associated protein 2.4; Keratin-associated protein 2-3; Keratin-associated protein 2-4; KRTAP2.3; KRTAP2.4; KRTAP2-3; KRTAP2-4 | |
| Human recombinant protein fragment corresponding to amino acids 1-128 of human KRTAP2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction