missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KSHV K8a Monoclonal Antibody (8C12G10G1)

Description
MA5-14768 targets KSHV K8a in indirect ELISA, IHC and WB applications and shows reactivity with Virus samples. The MA5-14768 immunogen is purified recombinant fragment of KSHV K8a expressed in E. Coli.
Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. KSHV K8 and K8.1 open reading frames are juxtaposed and span from nucleotide (nt) 74850 to 76695 of the virus genome. A K8 pre-mRNA overlaps the entire K8.1 coding region, and alternative splicing of KSHV K8 and K8.1 pre-mRNAs each produces three isoforms (alpha, beta and gamma) of the mRNAs.
Specifications
Specifications
| Antigen | KSHV K8a |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8C12G10G1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene Alias | Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of KSHV K8 alpha expressed in E. Coli. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?