missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ku70 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583871

Product Code. 16350825

  • € 582.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PPYFVALVPQ EEELDDQKIQ VTPPGFQLVF LPFADDKRKM PFTEKIMATP EQVGKMKAIV EKLRFTYRSD SFENPVLQQH FRNLEALALD LMEPEQAVDL Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 85%.

XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ku70
Polyclonal
Unconjugated
XRCC6
5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 70kDa; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CT; CTA-216E10.7; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTCBF; DNA repair protein XRCC6; G22p1; Ku autoantigen p70 subunit; ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Kup70; Lupus Ku autoantigen protein p70; ML8; OTTHUMP00000028581; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; XELAEV_18023498mg; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair complementing defective repair in Chinese hamster cells 6 L homeolog; X-ray repair cross complementing 6; X-ray repair cross-complementing protein 6; xrcc6; xrcc6.L
Rabbit
Antigen affinity chromatography
RUO
2547
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P12956
XRCC6
Recombinant protein corresponding to Human XRCC6. Recombinant protein control fragment (Product #RP-103007).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.