missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ku80/XRCC5 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-508JF646
This item is not returnable.
View return policy
Description
Ku80/XRCC5 Polyclonal antibody specifically detects Ku80/XRCC5 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot
Specifications
| Ku80/XRCC5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Hamster | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster | |
| A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein. | |
| 0.1 mL | |
| Cancer, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Stem Cell Markers | |
| 7520 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction