missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ku80/XRCC5 Antibody (XRCC5/8703R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21134F
This item is not returnable.
View return policy
Description
Ku80/XRCC5 Monoclonal antibody specifically detects Ku80/XRCC5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Ku80/XRCC5 | |
| Monoclonal | |
| FITC | |
| 80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster | |
| Recombinant fragment (around aa300-500) of human Ku80/XRCC5 (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Phospho Specific, Stem Cell Markers | |
| 7520 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| XRCC5/8703R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction