Learn More
Kudoa thyrsites Monoclonal Antibody (IPA-2F4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170114
Additional Details : Weight : 0.01000kg
Description
MA1-70114 detects Kudoa Thrysites in Kudoa thrysites samples. MA1-70114 has been successfully used in Western blot, immunofluorescence and flow cytometry procedures. The MA1-70114 immunogen is PercollTM-purified K. thyrsites spores.
Kudoa thrysites is a myxosporean parasite of marine fishes. It has a worldwide distribution, and infects a wide range of host species. This parasite is responsible for causing economic losses to the fisheries sector, by causing post-mortem "e;myoliquefaction"e;, a softening of the flesh to such an extent that the fish becomes unmarketable. It is not infective to humans.Specifications
Kudoa thyrsites | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Protozoa | |
Antibody | |
IgM |
Flow Cytometry, Immunocytochemistry, Western Blot | |
IPA-2F4 | |
Unconjugated | |
myxosporean | |
PercollTM-purified K. thyrsites spores. | |
250 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |