missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kuromanin chloride, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
Kuromanin (chloride), extracted from mulberry leaves, has been shown to improve blood glucose concentrations and lipid homeostasis and to reduce obesity.
Specifications
Specifications
| Chemical Name or Material | Kuromanin chloride |
| CAS | 7084-24-4 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C21H21ClO11 |
| Quantity | 10 mg |
| Synonym | Chrysontemin Cyanidin 3-O-glucoside chloride |
| Solubility Information | DMSO : 25 mg/mL (51.56 mM; Need ultrasonic) |
| SMILES | O[C@H]([C@@H](O)[C@@H]1O)[C@@H](O[C@@H]1CO)OC(C(C2=CC(O)=C(O)C=C2)=[O+]C3=CC(O)=C4)=CC3=C4O.[Cl-] |
| Molecular Weight (g/mol) | 484.84 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction