missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv11.3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06083-100ul
This item is not returnable.
View return policy
Description
Kv11.3 Polyclonal antibody specifically detects Kv11.3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kv11.3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Eag-related protein 3, ERG-3, ERG3eag related protein 3, ether-a-go-go related gene potassium channel 3, Ether-a-go-go-related gene potassium channel 3, Ether-a-go-go-related protein 3, hERG-3, HERG3erg3, Kv11.3MGC45986, potassium channel subunit HERG-3, potassium voltage-gated channel subfamily H member 7, potassium voltage-gated channel, subfamily H (eag-related), member 7, Voltage-gated potassium channel subunit Kv11.3 | |
| Produced in rabbits immunized with E. coli-derived Human Kv11.3 fragment. | |
| 100 μg | |
| Neuroscience | |
| 90134 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction