missing translation for 'onlineSavingsMsg'
Learn More

Kv3.2 Antibody (S410-17), Novus Biologicals™

Product Code. p-200081001 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18204734 100 μg 100µg
18685607 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18204734 Supplier Novus Biologicals Supplier No. NBP259328

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kv3.2 Monoclonal specifically detects Kv3.2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kv3.2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S410-17
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
Gene Alias Kv3.2, MGC138196, potassium voltage-gated channel subfamily C member 2, potassium voltage-gated channel, Shaw-related subfamily, member 2, Voltage-gated potassium channel Kv3.2
Gene Symbols KCNC2
Host Species Mouse
Immunogen Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3747
Test Specificity Detects 80-100kDa. Varies due to post-translational modifications.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.