Learn More
Kv4.2 Antibody (5B11B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52497
 View more versions of this product
Product Code. 18117699
  • € 552.00 / 0.10mg
Description
Kv4.2 Monoclonal specifically detects Kv4.2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kv4.2
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel
Mouse
70.5 kDa
0.1 mg
Neuronal Cell Markers, Neuroscience, Neurotransmission
3751
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
5B11B9
Unconjugated
PBS with 0.05% Sodium Azide
KCND2
Purified recombinant fragment of human KCND2 (AA: 27-184) expressed in E. Coli.
Protein A or G purified