Learn More
Novus Biologicals™ Kv4.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30825-25ug
This item is not returnable.
View return policy
Description
Kv4.2 Polyclonal antibody specifically detects Kv4.2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Kv4.2 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
| Synthetic peptide derived from the rat Kv4.2 conjugated to KLH | |
| 25 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3751 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |