missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv6.1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99150B
This item is not returnable.
View return policy
Description
Kv6.1 Polyclonal antibody specifically detects Kv6.1 in Human samples. It is validated for Western Blot
Specifications
| Kv6.1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| KCNG, KH2, kH2K13, KV6.1, MGC12878, potassium channel KH2, potassium channel Kv6.1, potassium voltage-gated channel subfamily G member 1, potassium voltage-gated channel, subfamily G, member 1, Voltage-gated potassium channel subunit Kv6.1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Kv6.1. | |
| 0.1 mL | |
| Neuroscience | |
| 3755 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction