Learn More
KvBeta1 Antibody (OTI4E7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71344AF647
Product Code. 18129722
  • € 751.00 / 0.10mL
Description
KvBeta1 Monoclonal specifically detects KvBeta1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KvBeta1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
KCNAB1
Full length recombinant protein of human KvBeta1 (NP_751891) produced in HEK293T cells.
0.1 mL
7881.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI4E7
Western Blot, Immunohistochemistry
AKR6A3, hKvb3, hKvBeta3, K(+) channel subunit beta-1, KCNA1Bpotassium channel beta 3 chain, Kvb1.3, KV-BETA-1, potassium channel beta3 subunit, potassium channel shaker chain beta 1a, potassium voltage-gated channel beta subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 1, voltage-gated potassium channel beta-1 subunit, voltage-gated potassium channel subunit beta-1
Mouse
Affinity Purified
Primary
Human, Mouse, Rat