missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KVbeta2 (KCNAB2) Polyclonal Antibody

KVbeta2 (KCNAB2) Polyclonal Antibody
Brand: Invitrogen PA568935
This item is not returnable.
View return policy
Description
KVbeta2 (KCNAB2) Polyclonal Antibody for Western Blot, IHC
Specifications
| KVbeta2 (KCNAB2) | |
| Polyclonal | |
| Unconjugated | |
| KCNAB2 | |
| AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; K(+) channel subunit beta-2; KCNA2B; KCNAB2; KCNAB2.1; KCNAB2.2; Kcnb3; KCNK2; KVB2; Kvbeta2; Kv-beta-2; Kvbeta2.1; Kvbeta2.2; neuroimmune protein F5; potassium channel; potassium channel Kv beta 2.1 subunit; potassium channel Kv beta 2.2 subunit; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel subfamily A regulatory beta subunit 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 1; potassium voltage-gated channel, shaker-related subfamily, beta member 2, transcript variant 2; voltage-gated potassium channel subunit beta-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16498, 8514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62482, Q13303 | |
| KCNAB2 | |
| synthetic peptide directed towards the middle region of human KCNAB2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction