missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KYNU Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583205
This item is not returnable.
View return policy
Description
Immunogen sequence: YAIESQLQLH GLNIEESMRM IKPREGEETL RIEDILEVIE KEGDSIAVIL FSGVHFYTGQ HFNIPAITKA GQAKGCYVGF DLAHAVGNVE LYLHDWGVDF ACWCSYKYLN AGAGGIAGAF IHEKHAHTIK PALVGWFGHE LSTRFK Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 89%.
Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene. Kynureninase is a pyridoxal-5'-phosphate (pyridoxal-P) dependent enzyme that catalyzes the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. Kynureninase is involved in the biosynthesis of NAD cofactors from tryptophan through the kynurenine pathway. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| KYNU | |
| Polyclonal | |
| Unconjugated | |
| Kynu | |
| 4432411A05Rik; HAMAP-Rule:MF_03017}; KYNU; kynureninase; kynureninase (L-kynurenine hydrolase); kynureninase {ECO:0000255; kynureninase-like protein; KYNUU; L-kynurenine hydrolase; L-kynurenine hydrolase {ECO:0000255 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16719 | |
| Kynu | |
| Recombinant protein corresponding to Human KYNU. Recombinant protein control fragment (Product #RP-89606). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction