missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kynureninase Antibody (004), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90525AF532
This item is not returnable.
View return policy
Description
Kynureninase Monoclonal specifically detects Kynureninase in Mouse samples. It is validated for Western Blot.
Specifications
| Kynureninase | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8942 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 4 | |
| Western Blot | |
| EC 3.7.1.3, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Kynureninase (Q9CXF0; Met1-Ser464). | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction