Learn More
L-Alanine Benzyl Ester Benzenesulfonic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | L-Alanine Benzyl Ester Benzenesulfonic Acid Salt |
| CAS | 101399-43-3 |
| Molecular Formula | C10 H13 N O2 . C6 H6 O3 S |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C10H13NO2.C6H6O3S/c1-8(11)10(12)13-7-9-5-3-2-4-6-9;7-10(8,9)6-4-2-1-3-5-6/h2-6,8H,7,11H2,1H3;1-5H,(H,7,8,9)/t8-;/m0./s1 |
| SMILES | C[C@H](N)C(=O)OCc1ccccc1.OS(=O)(=O)c2ccccc2 |
| IUPAC Name | benzenesulfonic acid;benzyl (2S)-2-aminopropanoate |
| Molecular Weight (g/mol) | 337.39 |
| Formula Weight | 337.0984 |